Learn More
AdipoGen IBMX,Chemical. CAS: 28822-58-4;
SDP
Supplier:  AdipoGen AGCR13512G001
Encompass_Preferred
IBMX, AdipoGen Life Sciences. Chemical. CAS: 28822-58-4. Formula: C10H14N4O2. MW: 222.3. Cell permeable, competitive, non-specific cAMP and cGMP phosphodiesterase inhibitor. Increases cAMP levels that activate PKA, leading to decreased proliferation, increased differentiation and induction of apoptosis. Enhances differentiation of 3T3-L1 cells. Non-selective adenosine receptor antagonist. Inhibits Ca2+ ion channels. Activates TNF-alpha. Adipogenic. Activates leukotriene synthesis. Reduces inflammation and innate immunity.
Catalog No. 50-464-728