Learn More
BIOGEMS INTERNATIONAL INC IBMX 100mg
SDP
Catalog No. 501529991
Encompass
IBMX is a general inhibitor of cyclic nucleotide phosphodiesterases, increasing the level of intracellular cAMP. It is used to increase cell differentiation, decrease proliferation, and induce apoptosis. IBMX is reported to inhibit TNF-alpha, reduce inflammation, and activate PKA.
Catalog No. 50-152-9991 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 2885842100MG
IBMX is a general inhibitor of cyclic nucleotide phosphodiesterases, increasing the level of intracellular cAMP. It is used to increase cell differentiation, decrease proliferation, and induce apoptosis. IBMX is reported to inhibit TNF-alpha, reduce inflammation, and activate PKA.