Learn More
TARGETMOL CHEMICALS INC IBMX 200MG
SDP
Catalog No. 502879389
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. IBMX (Methylisobutylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor with inhibitory activity against PDE3, PDE4, and PDE5 (IC50=6.5/26.3/31.7 uM). IBMX enhances the intracellular cAMP level. Purity 99.8%
Catalog No. 50-287-9389 Supplier TARGETMOL CHEMICALS INC Supplier No. T1713200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. IBMX (Methylisobutylxanthine) is a broad-spectrum phosphodiesterase (PDE) inhibitor with inhibitory activity against PDE3, PDE4, and PDE5 (IC50=6.5/26.3/31.7 uM). IBMX enhances the intracellular cAMP level. Purity 99.8%