Learn More
Selleck Chemical LLC IBMX
SDP
Catalog No. 5000003629
Encompass_Preferred
IBMX (Isobutylmethylxanthine 1-Methyl-3-Isobutylxanthine) is a nonspecific inhibitor of phosphodiesterase (PDE) with IC50 values of 6 5 1 2 26 3 3 9 and 31 7 5 3 M for PDE3 4 and 5 respectively It may enhance the intracellular cAMP levels and also acts as an adenosine (A1) receptor antagonist
Catalog No. 50-000-03629 Supplier Selleck Chemical LLC Supplier No. S583610MM/1ML
IBMX (Isobutylmethylxanthine 1-Methyl-3-Isobutylxanthine) is a nonspecific inhibitor of phosphodiesterase (PDE) with IC50 values of 6 5 1 2 26 3 3 9 and 31 7 5 3 M for PDE3 4 and 5 respectively It may enhance the intracellular cAMP levels and also acts as an adenosine (A1) receptor antagonist