Learn More
Selleck Chemical LLC IBMX 5mg 28822-58-4 Isobutylmethylxanthine, 1-Methyl-3-Isobutylxanthine
SDP
Supplier:  Selleck Chemical LLC S58365MG
Encompass_Preferred
IBMX (Isobutylmethylxanthine, 1-Methyl-3-Isobutylxanthine) is a nonspecific inhibitor of phosphodiesterase (PDE) with IC50 values of 6.5¿1.2, 26.3¿3.9 and 31.7¿5.3 ?M for PDE3, 4 and 5 respectively. It may enhance the intracellular cAMP levels and also acts as an adenosine (A1) receptor antagonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8425