Learn More
Abcam IBM x , Non-specific cAMP and cGMP inhibitor, 100MG
Catalog No. 502584566
Encompass_Preferred
MW 222.24 Da, Purity >98%. Non-specific cAMP and cGMP inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4566 Supplier Abcam Supplier No. AB120840100MG
MW 222.24 Da, Purity >98%. Non-specific cAMP and cGMP inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement