Learn More
Cayman Chemical IbotenIc AcId 10mg
SDP
Catalog No. 502973012
Encompass_Preferred
A neuroexcitatory amino acid that functions as a NMDA and metabotropic glutamate receptor agonist; used to induce brain lesions in animal models of cognitive dysfunction
Catalog No. 50-297-3012 Supplier Cayman Chemical Supplier No. 1458410 MG
A neuroexcitatory amino acid that functions as a NMDA and metabotropic glutamate receptor agonist; used to induce brain lesions in animal models of cognitive dysfunction