Learn More
TARGETMOL CHEMICALS INC Ibotenic acid 10MG
SDP
Catalog No. 502877299
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ibotenic acid (RS-42358-197) has agonist activity at both the NMDA and trans-ACPD or metabotropic quisqualate receptors. Purity 99.7%
Catalog No. 50-287-7299 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ027910MG
Add to Cart
Add to Cart
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ibotenic acid (RS-42358-197) has agonist activity at both the NMDA and trans-ACPD or metabotropic quisqualate receptors. Purity 99.7%