Learn More
Enzo Life Sciences Ibotenic acid (1mg). CAS: 2552-55-8
SDP
Supplier:  Enzo Life Sciences BMLEA1200001
Encompass_Preferred
Potent glutamate receptor agonist. Displays full agonist activity at mGlu1 and mGlu5. Alternative name: alpha-Amino-3-hydroxy-5-isoxazoleacetic acid. Purity: ≥98% (HPLC). Solubility: Soluble in water (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0528