Learn More
Apexbio Technology LLC Ibotenic acid(Synonyms: Ibotenate, Ibotenic acid hydrate, α-Amino-3-hydroxy-5-isoxazoleacetic acid, 3-Isoxazoleacetic acid), 25mg, CAS: 2552-55-8.
SDP
Supplier:  Apexbio Technology LLC B624625
Encompass_Preferred
Ibotenic acid (CAS 2552-55-8) is a small-molecule agonist targeting N-methyl-D-aspartate (NMDA) and metabotropic glutamate receptors Through receptor-mediated modulation of glutamatergic signaling pathways ibotenic acid induces alterations in neuronal activity In neuroscience research it is commonly utilized to establish animal models of neurodegenerative disorders and to investigate underlying neural mechanisms of disease states
Catalog No. 50-000-61065