Learn More
Medchemexpress LLC HY-N2311 5mg Medchemexpress, Ibotenic acid CAS:2552-55-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23115MG
Encompass_Preferred
Medchemexpress, HY-N2311 5mg Ibotenic acid CAS:2552-55-8 Ibotenic acid has agonist activity at both the N-methyl-D-aspartate ( NMDA ) and trans-ACPD or metabolotropic quisqualate (Q m ) receptor sites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3909