Learn More
Cayman Chemical Ibotenic Acid, Purity: ≥98%, CAS: 2552-55-8, 5 mg
SDP
Catalog No. 502972870
Encompass_Preferred
A neuroexcitatory amino acid that functions as a NMDA and metabotropic glutamate receptor agonist; used to induce brain lesions in animal models of cognitive dysfunction
Catalog No. 50-297-2870 Supplier Cayman Chemical Supplier No. 145845 MG
A neuroexcitatory amino acid that functions as a NMDA and metabotropic glutamate receptor agonist; used to induce brain lesions in animal models of cognitive dysfunction