Learn More
Medchemexpress LLC HY-117549 10mg Medchemexpress, Ibrolipim CAS:133208-93-2 Purity:>98%
SDP
Catalog No. 501969478
Encompass_Preferred
Medchemexpress, HY-117549 10mg Ibrolipim CAS:133208-93-2 Ibrolipim (NO-1886) is an orally active lipoprotein lipase (LPL)-promoting agent. Ibrolipim decreases plasma triglycerides, increases high-density lipoprotein cholesterol levels. Ibrolipim has renoprotective and hypolipidemic effects[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9478 Supplier Medchemexpress LLC Supplier No. HY11754910MG
Medchemexpress, HY-117549 10mg Ibrolipim CAS:133208-93-2 Ibrolipim (NO-1886) is an orally active lipoprotein lipase (LPL)-promoting agent. Ibrolipim decreases plasma triglycerides, increases high-density lipoprotein cholesterol levels. Ibrolipim has renoprotective and hypolipidemic effects[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.