Learn More
Medchemexpress LLC HY-10997 100mg , Ibrutinib CAS:936563-96-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10997100MG
Encompass_Preferred
HY-10997 100mg Ibrutinib CAS: 936563-96-1 Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9225