Learn More
Medchemexpress LLC HY-10997 200mg Medchemexpress, Ibrutinib CAS:936563-96-1 Purity:>98%
SDP
Catalog No. 501968729
Encompass_Preferred
Medchemexpress, HY-10997 200mg Ibrutinib CAS:936563-96-1 Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8729 Supplier Medchemexpress LLC Supplier No. HY10997200MG
Medchemexpress, HY-10997 200mg Ibrutinib CAS:936563-96-1 Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.