Learn More
Medchemexpress LLC HY-10997 50mg Medchemexpress, Ibrutinib CAS:936563-96-1 Purity:>98%
SDP
Catalog No. 502028803
Encompass_Preferred
Medchemexpress, HY-10997 50mg Ibrutinib CAS:936563-96-1 Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8803 Supplier Medchemexpress LLC Supplier No. HY1099750MG
Medchemexpress, HY-10997 50mg Ibrutinib CAS:936563-96-1 Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.