Learn More
AdipoGen Ibrutinib (5 mg)
SDP
Supplier:  AdipoGen AGCR13620M005
Encompass_Preferred
Chemical. CAS: 936563-96-1. Formula: C25H24N6O2. MW: 440.5. Potent and highly selective Bruton's tyrosine kinase (BTK) inhibitor. Orally bioavailable and irreversible inhibitor of BTK (IC50 = 0.5 nM) with antineoplastic activity. Binds to and irreversibly inhibits BTK activity, preventing B cell activation and B cell-mediated signaling. This leads to an inhibition of the growth of malignant B cells that overexpress BTK. Shown to inhibit autophosphorylation of BTK (IC50 = 11nM), phosphorylation of PLCgamma (IC50 = 29nM), a substrate of BTK and phosphorylation of ERK (IC50 = 13nM).
Catalog No. 50-205-3988