Learn More
Medchemexpress LLC Ibrutinib-biotin | 1599432-18-4 | MFCD30496724 | 99.7% | 1097.37 | C56H80N12O9S | 25 MG
SDP
Catalog No. 5000214601
Encompass_Preferred
Ibrutinib-biotin is a biotinylated probe in which the BTK inhibitor ibrutinib is linked to biotin via a long-chain linker (derived from patent WO2014059368A1 Compound 1-5). It is intended for research use to study Bruton's tyrosine kinase (BTK) binding, target engagement, and affinity-based assays.
  • Biotinylated probe for affinity capture and target engagement studies.
  • High potency against BTK with IC50 of 0.755-1.02 nM.
  • High purity (99.66%) suitable for biochemical and cellular assays.
  • Available in small quantities for assay development (25 mg).
  • Includes downloadable datasheet, COA, and SDS for experimental reproducibility.
Catalog No. 50-002-14601 Supplier Medchemexpress LLC Supplier No. HY10034225MG
Ibrutinib-biotin is a biotinylated probe in which the BTK inhibitor ibrutinib is linked to biotin via a long-chain linker (derived from patent WO2014059368A1 Compound 1-5). It is intended for research use to study Bruton's tyrosine kinase (BTK) binding, target engagement, and affinity-based assays.
  • Biotinylated probe for affinity capture and target engagement studies.
  • High potency against BTK with IC50 of 0.755-1.02 nM.
  • High purity (99.66%) suitable for biochemical and cellular assays.
  • Available in small quantities for assay development (25 mg).
  • Includes downloadable datasheet, COA, and SDS for experimental reproducibility.