Learn More
Cayman Chemical IbrutInIb-d5 500ug
SDP
Catalog No. 502986764
Encompass_Preferred
An internal standard for the quantification of ibrutinib by GC- or LC-MS
Catalog No. 50-298-6764 Supplier Cayman Chemical Supplier No. 22561500 UG
An internal standard for the quantification of ibrutinib by GC- or LC-MS