Learn More
Medchemexpress LLC HY-10997S 5mg Medchemexpress, Ibrutinib D5 CAS:1553977-17-5 Purity:>98%
SDP
Catalog No. 501968732
Encompass_Preferred
Medchemexpress, HY-10997S 5mg Ibrutinib D5 CAS:1553977-17-5 Ibrutinib D5 (PCI-32765 D5) is a deuterium labeled Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8732 Supplier Medchemexpress LLC Supplier No. HY10997S5MG
Medchemexpress, HY-10997S 5mg Ibrutinib D5 CAS:1553977-17-5 Ibrutinib D5 (PCI-32765 D5) is a deuterium labeled Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.