Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ibrutinib-mpea | 1710768-30-1 | MFCD31813735 | 99.7% | 581.71 | C32H39N9O2 | 10 MG
SDP

Catalog No. 5000299269
Encompass_Preferred

Ibrutinib-MPEA is a synthetic derivative of ibrutinib used as a research reagent to probe Bruton's tyrosine kinase (BTK) biology and structure-activity relationships. It is supplied as a high-purity solid suitable for biochemical and cellular assay applications.

  • High purity: 99.7%.
  • Molecular weight: 581.71 g/mol.
  • Molecular formula: C32H39N9O2.
  • Supplied as a 10 mg vial for laboratory use.
  • For research use only; not for human or veterinary applications.

Catalog No. 50-002-99269 Supplier Medchemexpress LLC Supplier No. HY4352110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ibrutinib-MPEA is a synthetic derivative of ibrutinib used as a research reagent to probe Bruton's tyrosine kinase (BTK) biology and structure-activity relationships. It is supplied as a high-purity solid suitable for biochemical and cellular assay applications.

  • High purity: 99.7%.
  • Molecular weight: 581.71 g/mol.
  • Molecular formula: C32H39N9O2.
  • Supplied as a 10 mg vial for laboratory use.
  • For research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.