Learn More
Medchemexpress LLC HY-43521 5mg Medchemexpress, Ibrutinib-MPEA CAS:1710768-30-1 Purity:>98%
SDP
Catalog No. 501970058
Encompass_Preferred
Medchemexpress, HY-43521 5mg Ibrutinib-MPEA CAS:1710768-30-1 Ibrutinib-MPEA (Compound 20) is ibrutinib derivative. Ibrutinib is a covalent and irreversible inhibitor of Bruton's tyrosine kinase (BTK) that has been used to treat haematological malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0058 Supplier Medchemexpress LLC Supplier No. HY435215MG
Medchemexpress, HY-43521 5mg Ibrutinib-MPEA CAS:1710768-30-1 Ibrutinib-MPEA (Compound 20) is ibrutinib derivative. Ibrutinib is a covalent and irreversible inhibitor of Bruton's tyrosine kinase (BTK) that has been used to treat haematological malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.