Learn More
Medchemexpress LLC HY-10997A 5mg Medchemexpress, Ibrutinib Racemate CAS:936563-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10997A5MG
Encompass_Preferred
Medchemexpress, HY-10997A 5mg Ibrutinib Racemate CAS:936563-87-0 Ibrutinib Racemate (PCI-32765 Racemate) is the racemate of Ibrutinib. Ibrutinib is a selective, irreversible Btk inhibitor with IC50 value of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8730