Learn More
Bioworld IBS/Casein, 200 mL
Supplier:  Bioworld 207200031
Encompass_Preferred
IBS/Casein All blocks and diluents are supplied in a 10X concentration of physiological saline buffer.Sold as 2 x 100mL or 500mL.Technical Information: pH range: 7.0 (+/- 0.1) based upon an Imidazole physiologic buffer
Catalog No. 50-198-402