Learn More
Bioworld IBS/Fish Gelatin (10X), 200 mL
Supplier:  Bioworld 207200041
Encompass_Preferred
IBS/Fish Gelatin IBS Buffered fish gelatin 5% blocks, diluents and coating solutions are supplied as 10X concentrates.Sold as 2 x 100mL. Technical Information: pH range: 7.0 (+/- 0.1) based upon an Imidazole physiologic saline buffer
Catalog No. 50-198-404