Learn More
Aviva Systems Biology Corp IBSP Antibody - N-terminal region (ARP36681_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36681_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IBSP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IBSP. Alternate names: BSP, BNSP, SP-II, BSP-IIApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-53057