Learn More
Medchemexpress LLC IBSP Human His-SUM 20ug
SDP
Catalog No. 5000182599
Encompass_Preferred
The IBSP protein has a strong affinity for hydroxyapatite and plays a crucial role in mineralized matrix formation indicating its overall nature in this regard Its tight binding suggests an important role in cell-matrix interactions particularly in promoting RGD-dependent cell attachment IBSP Protein Human (His-SUMO) is the recombinant human-derived IBSP protein expressed by E coli with N-6 His N-SUMO labeled tag
Catalog No. 50-001-82599 Supplier Medchemexpress LLC Supplier No. HYP7224120UG
The IBSP protein has a strong affinity for hydroxyapatite and plays a crucial role in mineralized matrix formation indicating its overall nature in this regard Its tight binding suggests an important role in cell-matrix interactions particularly in promoting RGD-dependent cell attachment IBSP Protein Human (His-SUMO) is the recombinant human-derived IBSP protein expressed by E coli with N-6 His N-SUMO labeled tag