Learn More
Formlabs Inc IBT Flex Resin (Form 3) 1 L
SDP
Catalog No. NC3782418
Encompass
IBT Flex Resin is a Class I biocompatible material with enhanced flexibility, strength, translucency, and color to guarantee optimal clinical outcomes while providing a great patient experience and for seamless and precise transfer of orthodontic brackets and restorative composite materials. 3D print flexible and tear-resistant translucent trays and guides that save you time and deliver consistent, predictable outcomes.
Catalog No. NC3782418 Supplier Formlabs Inc Supplier No. RSF2IBFL01
IBT Flex Resin is a Class I biocompatible material with enhanced flexibility, strength, translucency, and color to guarantee optimal clinical outcomes while providing a great patient experience and for seamless and precise transfer of orthodontic brackets and restorative composite materials. 3D print flexible and tear-resistant translucent trays and guides that save you time and deliver consistent, predictable outcomes.