Learn More
Ibt Bioservices IBT Marburg-Musoke virus-like particles (MMARV VLP), Cat # 0566-001, Marburg Virus, Marburg VLP, Musoke, Column chromatography, Western Blot, Liquid, Storage: -80°C
SDP
Catalog No. NC3958523
Encompass_Preferred
Virus-like Particles (VLP’s) expressing recombinant Sudan virus (SUDV) glycoprotein (GP), nucleoprotein (NP), and matrix protein (VP40). These VLP’s are produced in Sf9 insect cells through infection with a recombinant baculovirus. Since these VLP’s mimic Sudan virus but do not contain genetic material, thus are not infectious and make an ideal candidate as a vaccine and also as a tool to enhance filovirus research.Supplied in PBS (supplemented with glycerol, arginine and glutamic acid). Store at 2-3 weeks at -20°C, -80°C long term
Catalog No. NC3958523 Supplier Ibt Bioservices Supplier No. 0566001
Virus-like Particles (VLP’s) expressing recombinant Sudan virus (SUDV) glycoprotein (GP), nucleoprotein (NP), and matrix protein (VP40). These VLP’s are produced in Sf9 insect cells through infection with a recombinant baculovirus. Since these VLP’s mimic Sudan virus but do not contain genetic material, thus are not infectious and make an ideal candidate as a vaccine and also as a tool to enhance filovirus research.Supplied in PBS (supplemented with glycerol, arginine and glutamic acid). Store at 2-3 weeks at -20°C, -80°C long term