Learn More
Ibt Bioservices IBT Mouse anti-Ebola Virus Glycoprotein mAb (4F3), Cat # 0201-020, Mouse, Monoclonal, Ebola Virus, Ebola glycoprotein, Zaire, Protein A/G, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Catalog No. NC3958477
Encompass_Preferred
Protein G purified mouse monoclonal antibody reactive to EBOV glycoprotein (GP). The antibody detects glycoprotein in virus-like particles (VLP) and EBOV glycoprotein without the transmembrane region (rGPΔTM).Supplied in PBS in frozen format with no preservatives added. Store at +4°C for 2-3 weeks; -80°C for long-term
Catalog No. NC3958477 Supplier Ibt Bioservices Supplier No. 0201020
Protein G purified mouse monoclonal antibody reactive to EBOV glycoprotein (GP). The antibody detects glycoprotein in virus-like particles (VLP) and EBOV glycoprotein without the transmembrane region (rGPΔTM).Supplied in PBS in frozen format with no preservatives added. Store at +4°C for 2-3 weeks; -80°C for long-term