Learn More
Ibt Bioservices IBT Recombinant Influenza HA (H5 Vietnam), Cat # 1501-001, Influenza A, Hemagglutinin from A/Vietnam/1203/2004 (H5N1), Column chromatography, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Catalog No. NC3958513
Encompass_Preferred
Recombinant, Influenza Hemagglutinin Full-Length Glycoprotein from virus strain A/Vietnam/1203/2004 (H5N1). Recombinant HA (rHA) is supplied as an affinity purified protein. rHA is produced in Sf9 insect cells using baculovirus for expression and purified by FPLC.Supplied in Tris buffered saline and 0.01% non-ionic surfactant. Store at -20°C for 2-3 weeks, -80°C for long term.
Catalog No. NC3958513 Supplier Ibt Bioservices Supplier No. 1501001
Recombinant, Influenza Hemagglutinin Full-Length Glycoprotein from virus strain A/Vietnam/1203/2004 (H5N1). Recombinant HA (rHA) is supplied as an affinity purified protein. rHA is produced in Sf9 insect cells using baculovirus for expression and purified by FPLC.Supplied in Tris buffered saline and 0.01% non-ionic surfactant. Store at -20°C for 2-3 weeks, -80°C for long term.