Learn More
Medchemexpress LLC IBT6A | 1022150-12-4 | 99.6% | 500 MG

Supplier: Medchemexpress LLC HY13036A500MG
IBT6A is identified as an impurity of Ibrutinib and can be utilized in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor. IBT6A can also be used in the synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs).
- Impurity of Ibrutinib
- Used in synthesis of Ibrutinib dimer and adduct
- Related to a selective, irreversible Btk inhibitor
- Utilized in synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs)
- Available as a white to yellow solid
- Molecular weight of 386.45
- Chemical formula is C22H22N6O
- Soluble in DMSO at 50 mg/mL (129.38 mM)
- Intended for research use only
- Long-term storage at -20°C for powder, -80°C for solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.