Learn More
Medchemexpress LLC IBT6A hydrochloride | 1553977-42-6 | 99.7% | 1 ML
SDP
Catalog No. 5000340456
Encompass_Preferred
IBT6A hydrochloride is an impurity of Ibrutinib. It is used in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is known as a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
  • Impurity of Ibrutinib.
  • Utilized in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct.
  • Can be used in the synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs).
  • Relevant to the study of Ibrutinib, a selective, irreversible Btk inhibitor.
Catalog No. 50-003-40456 Supplier Medchemexpress LLC Supplier No. HY13036B10MM/1ML
IBT6A hydrochloride is an impurity of Ibrutinib. It is used in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is known as a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
  • Impurity of Ibrutinib.
  • Utilized in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct.
  • Can be used in the synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs).
  • Relevant to the study of Ibrutinib, a selective, irreversible Btk inhibitor.