Learn More
Cayman Chemical IBTPIodIde 10mg
SDP
Catalog No. 502965587
Encompass_Preferred
A lipophilic cation that is accumulated in mitochondria and forms stable thioether adducts in a thiol-specific manner
Catalog No. 50-296-5587 Supplier Cayman Chemical Supplier No. 1880210 MG
A lipophilic cation that is accumulated in mitochondria and forms stable thioether adducts in a thiol-specific manner