Learn More
Hach Company IBU TNTplus Vial Test
Supplier:  Hach Company TNT817
Encompass
TNTplus Test International Bitter Units ( 2 IBU) For use with your Hach DR6000 spectrophotometer TNT817 offers you the ability to measure international Bitter Units with less hassle Additionally these vials provide high quality results free from potential external influencers They are designed to simplify your brewing analysis with accurate results every time
Catalog No. 50-002-07013