Learn More
TARGETMOL CHEMICALS INC Ibudilast 200mg
SDP
Catalog No. NC1624014
Encompass_Preferred
Ibudilast(KC-404;AV-411;MN-166) is a relatively nonselective phosphodiesterase inhibitor. It is approved for use as an anti-inflammatory in Japan.
Catalog No. NC1624014 Supplier TARGETMOL CHEMICALS INC Supplier No. T2137200MG
Ibudilast(KC-404;AV-411;MN-166) is a relatively nonselective phosphodiesterase inhibitor. It is approved for use as an anti-inflammatory in Japan.