Learn More
Medchemexpress LLC Ibufenac | 1553-60-2 | MFCD00864374 | 98.0% | 192.26 g/mol | C12H16O2 | 250 MG
SDP
Catalog No. 5000209285
Encompass_Preferred
Ibufenac is an orally active nonsteroidal anti-inflammatory agent with analgesic and antipyretic effects, used in research to evaluate anti-inflammatory activity and in animal models of rheumatic disease.
  • Orally active nonsteroidal anti-inflammatory agent.
  • Exerts anti-inflammatory, analgesic, and antipyretic effects.
  • Demonstrated activity in in vitro and in vivo inflammation models.
  • Available as solid and DMSO solution formats for research use.
  • High purity (98.01%) and defined physicochemical properties.
Catalog No. 50-002-09285 Supplier Medchemexpress LLC Supplier No. 250MGHYW040672
Ibufenac is an orally active nonsteroidal anti-inflammatory agent with analgesic and antipyretic effects, used in research to evaluate anti-inflammatory activity and in animal models of rheumatic disease.
  • Orally active nonsteroidal anti-inflammatory agent.
  • Exerts anti-inflammatory, analgesic, and antipyretic effects.
  • Demonstrated activity in in vitro and in vivo inflammation models.
  • Available as solid and DMSO solution formats for research use.
  • High purity (98.01%) and defined physicochemical properties.