Learn More
Selleck Chemical LLC Ibuprofen (NSC 256857) S1638-50mg
SDP
Catalog No. 503108488
Encompass_Preferred
Ibuprofen (NSC 256857 Dolgesic) is an anti-inflammatory inhibitor targeting COX-1 and COX-2 with IC50 of 13 M and 370 M respectively
Catalog No. 50-310-8488 Supplier Selleck Chemical LLC Supplier No. S163850MG
Ibuprofen (NSC 256857 Dolgesic) is an anti-inflammatory inhibitor targeting COX-1 and COX-2 with IC50 of 13 M and 370 M respectively