Learn More
Medchemexpress LLC HY-78131 1g , Ibuprofen CAS:15687-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY781311G
Encompass_Preferred
HY-78131 1g Ibuprofen CAS: 15687-27-1 Ibuprofen ((±)-Ibuprofen) is a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. Ibuprofen inhibits cell proliferation, angiogenesis, and induces cell apoptosis. Ibuprofen is a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. Ibuprofen ((±)-Ibuprofen) can be used in the research of pain, swelling, inflammation, infection, immunology, cancers[5][8]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1099