Learn More
Cayman Chemical (±)-Ibuprofen, Purity: ≥99%, CAS: 15687-27-1, 500 mg
SDP
Catalog No. 502896303
Encompass_Preferred
An NSAID and non-selective COX inhibitor (IC50s = 2.6 and 1.3 µM for human recombinant COX-1 and COX-2, respectively); inhibits late-phase formalin-induced paw licking in mice (ED50 = 6.1 mg/kg); inhibits acetic acid-induced writhing in mice (ED50 = 0.47 mg/kg)
Catalog No. 50-289-6303 Supplier Cayman Chemical Supplier No. 70280500 MG
An NSAID and non-selective COX inhibitor (IC50s = 2.6 and 1.3 µM for human recombinant COX-1 and COX-2, respectively); inhibits late-phase formalin-induced paw licking in mice (ED50 = 6.1 mg/kg); inhibits acetic acid-induced writhing in mice (ED50 = 0.47 mg/kg)