Learn More
Enzo Life Sciences Ibuprofen (5g). CAS: 15687-27-1
SDP
Supplier:  Enzo Life Sciences BMLEI1640005
Encompass_Preferred
Selective cyclooxygenase (COX) inhibitor (IC50=14.9 µM). Inhibits PGH synthase-1 and PGH synthase-2 with comparable potency (ID50=8.9-14 and 7.2-8.2 µM respectively; data are for (S) isomer). Clinically useful NSAID. Inhibits the neutrophil-dependent edema response of rabbit skin to C5a/PGE2. Alternative name: alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, (+/-)-2-(4-Isobutylphenyl)propionic acid. Purity: ≥99% (HPLC). Solubility: Soluble in ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0582