Learn More
Cayman Chemical Ibuprofen CarboxylIc ACD 500ug
SDP
Catalog No. 502934813
Encompass_Preferred
A major metabolite of ibuprofen; prevents fructose-, cyanate-, and prednisolone-induced inactivation of catalase and fructose- and cyanate-induced inactivation of fumarase at a concentration of 2 mM; has been found in bank filtrate and surface water, as well as influent and effluent wastewater, and is considered a micropollutant
Catalog No. 50-293-4813 Supplier Cayman Chemical Supplier No. 26594500 UG
A major metabolite of ibuprofen; prevents fructose-, cyanate-, and prednisolone-induced inactivation of catalase and fructose- and cyanate-induced inactivation of fumarase at a concentration of 2 mM; has been found in bank filtrate and surface water, as well as influent and effluent wastewater, and is considered a micropollutant