Learn More
Cayman Chemical Ibuprofen Carboxylic Acid, Purity: ≥98% (mixture of diastereomers), CAS: 15935-54-3, 1 mg
SDP
Catalog No. 502935467
Encompass_Preferred
A major metabolite of ibuprofen; prevents fructose-, cyanate-, and prednisolone-induced inactivation of catalase and fructose- and cyanate-induced inactivation of fumarase at a concentration of 2 mM; has been found in bank filtrate and surface water, as well as influent and effluent wastewater, and is considered a micropollutant
Catalog No. 50-293-5467 Supplier Cayman Chemical Supplier No. 265941 MG
A major metabolite of ibuprofen; prevents fructose-, cyanate-, and prednisolone-induced inactivation of catalase and fructose- and cyanate-induced inactivation of fumarase at a concentration of 2 mM; has been found in bank filtrate and surface water, as well as influent and effluent wastewater, and is considered a micropollutant