Learn More
Cayman Chemical (±)-Ibuprofen-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 121662-14-4, 10 mg
SDP
Catalog No. 502968308
Encompass_Preferred
An internal standard for the quantification of ibuprofen by GC- or LC-MS
Catalog No. 50-296-8308 Supplier Cayman Chemical Supplier No. 1825510 MG
An internal standard for the quantification of ibuprofen by GC- or LC-MS