Learn More
Medchemexpress LLC Ibuprofen-d3 | 121662-14-4 | 99.94% | C13H15D3O2 | 5 MG
SDP
Catalog No. 5000433157
Encompass_Preferred
Ibuprofen-d3 is a deuterium labeled Ibuprofen and functions as a COX-1 and COX-2 inhibitor. It is utilized as a tracer and internal standard for quantitative analysis in various analytical methods.
  • Functions as a COX-1 and COX-2 inhibitor
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for methods like NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-33157 Supplier Medchemexpress LLC Supplier No. HY78131S5MG
Ibuprofen-d3 is a deuterium labeled Ibuprofen and functions as a COX-1 and COX-2 inhibitor. It is utilized as a tracer and internal standard for quantitative analysis in various analytical methods.
  • Functions as a COX-1 and COX-2 inhibitor
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for methods like NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs