Learn More
Medchemexpress LLC Ibuprofen-d3 | 121662-14-4 | 99.94% | C13H15D3O2 | 50 MG
SDP
Catalog No. 5000434558
Encompass_Preferred
Ibuprofen-d3 is a deuterium labeled Ibuprofen, acting as a COX-1 and COX-2 inhibitor with IC50s of 13 μM and 370 μM. It incorporates stable heavy isotopes, such as deuterium, into drug molecules as tracers for quantitation during drug development. This process has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34558 Supplier Medchemexpress LLC Supplier No. HY78131S50MG
Ibuprofen-d3 is a deuterium labeled Ibuprofen, acting as a COX-1 and COX-2 inhibitor with IC50s of 13 μM and 370 μM. It incorporates stable heavy isotopes, such as deuterium, into drug molecules as tracers for quantitation during drug development. This process has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS