Learn More
Medchemexpress LLC Ibuprofen-d3 | 121662-14-4 | MFCD06658737 | 99.9% | 209.30 | C13H15D3O2 | 10 MG
SDP
Catalog No. 5000299275
Encompass_Preferred
Ibuprofen-d3 is the deuterium-labeled form of ibuprofen used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid and retains ibuprofen's COX-1 and COX-2 inhibitory profile, making it suitable for analytical and research applications.
  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High purity (≈99.9%) for reliable quantitation.
  • Soluble in DMSO up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored protected from light at recommended temperatures.
  • Available in small analytical sizes suitable for standards and assays.
Catalog No. 50-002-99275 Supplier Medchemexpress LLC Supplier No. HY78131S10MG
Ibuprofen-d3 is the deuterium-labeled form of ibuprofen used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid and retains ibuprofen's COX-1 and COX-2 inhibitory profile, making it suitable for analytical and research applications.
  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High purity (≈99.9%) for reliable quantitation.
  • Soluble in DMSO up to 100 mg/mL with ultrasonic assistance.
  • Stable when stored protected from light at recommended temperatures.
  • Available in small analytical sizes suitable for standards and assays.