Learn More
Medchemexpress LLC Ibuprofen-d3 sodium | 1219805-09-0 | MFCD11983518 | 99.0% | 231.28 | C13H14D3NaO2 | 10 MG
SDP
Catalog No. 5000298413
Encompass_Preferred
Ibuprofen-d3 sodium is the deuterium-labeled sodium salt of ibuprofen supplied as a solid research reagent. It is intended for use as an isotopic internal standard or tracer in analytical, pharmacokinetic, and metabolic studies, particularly LC-MS/MS methods. The material is supplied at high chemical purity and is for research use only.
  • Isotopically labeled internal standard for LC-MS quantitation.
  • High chemical purity (reported 99.0%).
  • Suitable for pharmacokinetic and metabolic tracing studies.
  • Solid powder, stable when stored sealed and away from moisture.
  • Available in small research pack sizes, e.g. 10 MG.
Catalog No. 50-002-98413 Supplier Medchemexpress LLC Supplier No. HY78131CS10MG
Ibuprofen-d3 sodium is the deuterium-labeled sodium salt of ibuprofen supplied as a solid research reagent. It is intended for use as an isotopic internal standard or tracer in analytical, pharmacokinetic, and metabolic studies, particularly LC-MS/MS methods. The material is supplied at high chemical purity and is for research use only.
  • Isotopically labeled internal standard for LC-MS quantitation.
  • High chemical purity (reported 99.0%).
  • Suitable for pharmacokinetic and metabolic tracing studies.
  • Solid powder, stable when stored sealed and away from moisture.
  • Available in small research pack sizes, e.g. 10 MG.