Learn More
Cayman Chemical (±)-Ibuprofen-d3 (sodium salt), Purity: ≥99% deuterated forms (d1-d3), CAS: 1219805-09-0, 50 mg
SDP
Catalog No. 502912071
Encompass_Preferred
An internal standard for the quantification of (±)-ibuprofen by GC- or LC-MS
Catalog No. 50-291-2071 Supplier Cayman Chemical Supplier No. 3633250 MG
An internal standard for the quantification of (±)-ibuprofen by GC- or LC-MS