Learn More
Cayman Chemical (±)-Ibuprofen-d3 (sodium salt), Purity: ≥99% deuterated forms (d1-d3), CAS: 1219805-09-0, 5 mg
SDP
Catalog No. 502912165
Encompass_Preferred
An internal standard for the quantification of (±)-ibuprofen by GC- or LC-MS
Catalog No. 50-291-2165 Supplier Cayman Chemical Supplier No. 363325 MG
An internal standard for the quantification of (±)-ibuprofen by GC- or LC-MS